Ethyl-2-(2-amino-4-thiazolyl)-2-(hydroxyimino)acetate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(2-Amino-4-thiazolyl)-2-(hydroxyimino)acetic Acid Ethyl Ester

OR936499-25G OR936499-5G
77425-412EA 131.42 USD
77425-412 77425-414
Ethyl-2-(2-amino-4-thiazolyl)-2-(hydroxyimino)acetate
Ethyl-2-(2-amino-4-thiazolyl)-2-(hydroxyimino)acetate
Formula: C₇H₉N₃O₃S
MW: 215.23 g/mol
Melting Pt: 195…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00010415
CAS Number: 60845-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR