1H,1H,5H-Octafluoropentyl 1,1,2,2-tetrafluoroethyl ether 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
2,2,3,3,4,4,5,5-Octafluoropentyl 1,1,2,2-Tetrafluoroethyl Ether, 1,1,2,2,3,3,4,4-Octafluoro-5-(1,1,2,2-tetrafluoroethoxy)pentane
PC10021-100G
PC10021-25G
77426-500EA
289.34
USD
77426-500
77426-502
1H,1H,5H-Octafluoropentyl 1,1,2,2-tetrafluoroethyl ether 97%
1H,1H,5H-Octafluoropentyl 1,1,2,2-tetrafluoroethyl ether
Formula:
C₇H₄F₁₂O MW: 332.09 g/mol Boiling Pt: 133 °C Storage Temperature: Ambient |
MDL Number:
MFCD00155904 CAS Number: 16627-71-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...