2-Fluoro-6-methoxyaniline

Supplier: APOLLO SCIENTIFIC
Warning

PC0872-1G PC0872-250MG PC0872-5G
77426-430EA 23.42 USD
77426-430 77426-432 77426-434
2-Fluoro-6-methoxyaniline
2-Fluoro-6-methoxyaniline
Formula: C₇H₈FNO
MW: 141.1429 g/mol
Boiling Pt: 208 °C/756 mmHg
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD03788538
CAS Number: 446-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR