3-Aminobutan-1-ol 97%
Supplier: APOLLO SCIENTIFIC
Synonyms:
3-Amino-butan-1-ol
OR912855-10G
OR912855-1G
OR912855-5G
77424-918EA
66.22
USD
77424-918
77424-920
77424-922
3-Aminobutan-1-ol 97%
3-Aminobutan-1-ol
Formula:
C₄H₁₁NO Storage Temperature: Ambient |
MDL Number:
MFCD03412694 CAS Number: 2867-59-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...