2-Methylpyrimidine-4-carbaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR904478-1G OR904478-250MG OR904478-5G
77424-768EA 183.38 USD
77424-768 77424-770 77424-772
2-Methylpyrimidine-4-carbaldehyde 98%
2-Methylpyrimidine-4-carbaldehyde
Formula: C₆H₆N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD09832936
CAS Number: 1004-17-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR