N-[3-(Aminomethyl)phenyl]-N-methylacetamide hydrochloride 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR7776-1G OR7776-5G
77424-324EA 192.56 USD
77424-324 77424-326
N-[3-(Aminomethyl)phenyl]-N-methylacetamide hydrochloride 95+%
N-[3-(Aminomethyl)phenyl]-N-methylacetamide hydrochloride
Formula: C₁₀H₁₅ClN₂O
MW: 214.7 g/mol
Melting Pt: 188…191 °C
Storage Temperature: Ambient
MDL Number: MFCD04039172
CAS Number: 849020-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR