3-Allylsalicylaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-AllylSalicylaldehyde, 3-Allyl-2-hydroxybenzaldehyde

OR7500T-25G OR7500T-5G
77424-224EA 470.7 USD
77424-224 77424-226
3-Allylsalicylaldehyde
3-Allylsalicylaldehyde

A synthetic intermediate for flavone synthesis.

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Melting Pt: 45 °C
Density: 1.099 g/cm³ (25 °C)
Flash Pt: 106.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00037364
CAS Number: 24019-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR