4-(Methylsulfonyl)-2-nitrobenzaldehyde 95+%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR7647-250MG
OR7647-1G
77424-256EA
41.78
USD
77424-256
77424-254
4-(Methylsulfonyl)-2-nitrobenzaldehyde 95+%
4-(Methylsulfonyl)-2-nitrobenzaldehyde
Formula:
C₈H₇NO₅S Storage Temperature: Ambient |
MDL Number:
MFCD04037929 CAS Number: 849035-66-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...