4-Bromo-1,5-dimethyl-1H-pyrazole 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Bromo-1,5-dimethylpyrazole

OR910220-10G OR910220-1G OR910220-5G
77424-856EA 344.36 USD
77424-856 77424-858 77424-860
4-Bromo-1,5-dimethyl-1H-pyrazole 95%
4-Bromo-1,5-dimethyl-1H-pyrazole
Formula: C₅H₇BrN₂
MW: 175.0265 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09759006
CAS Number: 5775-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR