4-Bromocinnamic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-(4-Bromophenyl)acrylic acid

OR6580-100G OR6580-1KG OR6580-500G
77423-852EA 124.3 USD
77423-852 77423-854 77423-856
4-Bromocinnamic acid 98%
4-Bromocinnamic acid
Formula: C₉H₇BrO₂
MW: 227.06 g/mol
Melting Pt: 260…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00004394
CAS Number: 1200-07-3
EINECS: 214-850-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR