Benzo[b]furan-5-boronic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR7149-1G OR7149-5G
77424-088EA 81.5 USD
77424-088 77424-090
Benzo[b]furan-5-boronic acid
Benzo[b]furan-5-boronic acid
Formula: C₈H₇BO₃
Melting Pt: 216…220 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038999
CAS Number: 331834-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR