L(+)-Lysine monohydrate 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: L-(+)-Lysine, Hydrate, (S)-(+)-2,6-Diaminohexanoic acid monohydrate, (S)-(+)-Lysine monohydrate, L-Lysine hydrate, (S)-(+)-2,6-Diaminocaproic acid monohydrate, H-Lys-OH monohydrate, K monohydrate, H-Lys-OH·H₂O, Lys monohydrate

OR52570-100G OR52570-1KG OR52570-500G
77422-714EA 69.28 USD
77422-714 77422-716 77422-718
L(+)-Lysine monohydrate 98%
L(+)-Lysine monohydrate
Formula: H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
MW: 164.2 g/mol
Melting Pt: 225
Storage Temperature: Ambient
MDL Number: MFCD00151035
CAS Number: 39665-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR