tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Bis-Boc-pyrazolocarboxamidine, 1-[N,N'-Bis(tert-butoxycarbonyl)carbamimidoyl]-1H-pyrazole, N,N'-Di-BOC-1H-pyrazole-1-carboxamidine, N,N'-Di-Boc-1H-pyrazole-1-carboximidamide, 1H-Pyrazole-1-carboxamidine, N,N'-Bis-BOC protected, N,N'-Bis(tert-butoxycarbonyl)-1H-pyrazole-1-carboximidamide

OR40453-25G OR40453-5G
77421-148EA 110.04 USD
77421-148 77421-150
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate 98%
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate
Formula: C₁₄H₂₂N₄O₄
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD01075122
CAS Number: 152120-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR