2-(3-Methoxyphenyl)piperidine hydrochloride

Supplier: APOLLO SCIENTIFIC

OR321505-1G OR321505-250MG
77419-998EA 645.92 USD
77419-998 77420-000
2-(3-Methoxyphenyl)piperidine hydrochloride
2-(3-Methoxyphenyl)piperidine hydrochloride
Formula: C₁₂H₁₇NO·ClH
MW: 227.73 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02178980
CAS Number: 1187172-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR