Dibenzothiophene-2-boronic acid 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR360153-1G OR360153-25G OR360153-5G
77420-590EA 21.34 USD
77420-590 77420-592 77420-594
Dibenzothiophene-2-boronic acid 99%
Dibenzothiophene-2-boronic acid
Formula: C₁₂H₉BO₂S
Storage Temperature: Freezer
MDL Number: MFCD01318982
CAS Number: 668983-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR