3,5,6-Trichlorosalicylic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR350376-100G OR350376-25G
77420-504EA 45.84 USD
77420-504 77420-506
3,5,6-Trichlorosalicylic acid 97%
3,5,6-Trichlorosalicylic acid
Formula: C₇H₃Cl₃O₃
Melting Pt: 209…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00075245
CAS Number: 40932-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR