2-Indanone 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
beta-Hydrindone, 2-Oxoindane, 1H-Inden-2(3H)-one
OR30508-100G
OR30508-25G
OR30508-5G
77417-652EA
84.56
USD
77417-652
77418-702
77418-704
2-Indanone 98%
2-Indanone
Formula:
C₉H₈O Melting Pt: 51…54 °C Flash Pt: 100 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003792 CAS Number: 615-13-4 EINECS: 210-410-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...