2,3-Dihydro-1H-inden-2-amine hydrochloride 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Aminoindan hydrochloride, 2-Indanylamine hydrochloride

OR30439-1G OR30439-25G OR30439-5G
77413-852EA 58.08 USD
77413-852 77413-854 77413-856
2,3-Dihydro-1H-inden-2-amine hydrochloride 98%
2,3-Dihydro-1H-inden-2-amine hydrochloride
Formula: C₉H₁₂ClN
Melting Pt: 243…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00012549
CAS Number: 2338-18-3
EINECS: 219-048-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR