2-Chloro-1-methyl-1H-indole-3-carbaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR30993-1G OR30993-250MG
77419-302EA 46.2 USD
77419-302 77419-304
2-Chloro-1-methyl-1H-indole-3-carbaldehyde
2-Chloro-1-methyl-1H-indole-3-carbaldehyde
Formula: C₁₀H₈ClNO
MW: 193.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02253186
CAS Number: 24279-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR