2-Indolinone

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Oxindole, Oxindole, Indolin-2-one, 2,3-Dihydro-2-indolone

OR30278-25G OR30278-5G
77406-896EA 23.28 USD
77406-896 77406-898
2-Indolinone
2-Indolinone
Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 313
Melting Pt: 123…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00005711
CAS Number: 59-48-3
EINECS: 200-429-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR