trans-3-(3-Pyridyl)acrylic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR30410-100G OR30410-250G OR30410-25G
77413-190EA 156.9 USD
77413-190 77413-294 77413-296
trans-3-(3-Pyridyl)acrylic acid 98%
trans-3-(3-Pyridyl)acrylic acid
Formula: C₈H₇NO₂
MW: 149.15 g/mol
Melting Pt: 232…235 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006410
CAS Number: 19337-97-4
EINECS: 214-424-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR