Ethyl-2-amino-4-chlorothieno[2,3-d]-pyrimidine-6-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR303984-1G OR303984-500MG
77411-660EA 113.1 USD
77411-660 77412-268
Ethyl-2-amino-4-chlorothieno[2,3-d]-pyrimidine-6-carboxylate
Ethyl-2-amino-4-chlorothieno[2,3-d]-pyrimidine-6-carboxylate
Formula: C₉H₈ClN₃O₂S
MW: 257.7 g/mol
Melting Pt: 253…255 °C
Storage Temperature: Ambient
MDL Number: MFCD18074287
CAS Number: 847560-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR