tert-Butyl Hydrogen Hexadecanedioate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 16-(tert-Butoxy)-16-oxohexadecanoic acid, Hexadecanedioic acid Mono-tert-butyl ester, Boc-C14-COOH

OR303207-10G OR303207-1G OR303207-5G
77407-938EA 144.68 USD
77407-938 77408-070 77408-158
tert-Butyl Hydrogen Hexadecanedioate 98%
tert-Butyl Hydrogen Hexadecanedioate
Formula: C₂₀H₃₈O₄
MW: 342.52 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23136039
CAS Number: 843666-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR