1-(2-Methoxyethyl)-1H-pyrazol-4-amine

Supplier: APOLLO SCIENTIFIC

OR302397-100MG OR302397-250MG
77405-402EA 64.18 USD
77405-402 77405-584
1-(2-Methoxyethyl)-1H-pyrazol-4-amine
1-(2-Methoxyethyl)-1H-pyrazol-4-amine
Formula: C₆H₁₁N₃O
MW: 141.171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09965625
CAS Number: 948570-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR