4-(4-Morpholinylcarbonyl)phenylboronic acid 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
4-(4-Morpholinylcarbonyl)benzeneboronic acid, (4-(Morpholine-4-carbonyl)phenyl)boronic acid
OR10568-1G
OR10568-25G
OR10568-5G
77402-788EA
114.1
USD
77402-788
77402-790
77402-792
4-(4-Morpholinylcarbonyl)phenylboronic acid 98%
4-(4-Morpholinylcarbonyl)phenylboronic acid
Formula:
C₁₁H₁₄BNO₄ Melting Pt: 126…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411952 CAS Number: 389621-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...