3-Bromo-4-fluoroaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC9908-100G PC9908-25G
77418-766EA 41.78 USD
77418-766 77418-768
3-Bromo-4-fluoroaniline 98%
3-Bromo-4-fluoroaniline
Formula: C₆H₅BrFN
MW: 190.01 g/mol
Boiling Pt: 124 °C/11 mmHg
Melting Pt: 29…36 °C
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD03788559
CAS Number: 656-64-4
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR