4-(Trifluoromethoxy)phenyl isocyanate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Isocyanato-4-(trifluoromethoxy)benzene

PC7439Q-1G PC7439Q-5G
77417-956EA 21.32 USD
77417-956 77417-958
4-(Trifluoromethoxy)phenyl isocyanate 98%
4-(Trifluoromethoxy)phenyl isocyanate

  • May form small amount of sediment.

Formula: C₈H₄F₃NO₂
Boiling Pt: 78 °C/22 mmHg
Melting Pt: –29 °C
Density: 1.346 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00035702
CAS Number: 35037-73-1
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR