Lithium bis(trifluoromethylsulfonyl)imide 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Lithium Bis(Trifluoromethanesulfonyl)Imide, Bis(trifluoromethane)sulfonimide lithium salt, Lithium bis(trifluoromethanesulphonimide), Lithium bis((trifluoromethyl)sulphonyl)amide, Lithium bis((trifluoromethyl)sulfonyl)amide, Lithium bis(trifluoromethanesulfonimide), Lithium Bis(Trifluoromethanesulphonyl)Imide, Bis(trifluoromethane)sulphonimide lithium salt

PC6834-100G PC6834-25G PC6834-50G
77417-552EA 103.92 USD
77417-552 77417-554 77417-556
Lithium bis(trifluoromethylsulfonyl)imide 99%
Lithium bis(trifluoromethylsulfonyl)imide
Formula: C₂F₆LiNO₄S₂
MW: 287.09 g/mol
Melting Pt: 234…238 °C
Storage Temperature: Ambient
MDL Number: MFCD00210017
CAS Number: 90076-65-6
EINECS: 415-300-0
UN: 2923
ADR: 8+6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR