Bis(2-methoxyethyl)aminosulfur trifluoride 50% in THF
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC6001-100G
PC6001-10G
PC6001-25G
77417-246EA
330.1
USD
77417-246
77417-248
77417-250
Bis(2-methoxyethyl)aminosulfur trifluoride 50% in THF
Bis(2-methoxyethyl)aminosulfur trifluoride
Deoxo-FluorR (Registered trademark of Air Products), 50% solution in THF. Stable alternative to DAST.
Formula:
C₆H₁₄F₃NO₂S MW: 221.24 g/mol Density: 1.02 g/cm³ (25 °C) Flash Pt: <10 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01321415 CAS Number: 202289-38-1 UN: 3286 ADR: 3+6.1+8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...