Toluene-α-sulfonyl fluoride 99%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC6222M-100G
PC6222M-250G
PC6222M-25G
77417-318EA
140.6
USD
77417-318
77417-320
77417-322
Toluene-α-sulfonyl fluoride 99%
Toluene-α-sulphonyl fluoride
Inhibitor of Serine proteases: Toxicity LD50 <500mg/kgAffords alkenes via reaction with carbonyl compounds.
Formula:
C₇H₇FO₂S Melting Pt: 92…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007424 CAS Number: 329-98-6 EINECS: 206-350-2 UN: 2928 ADR: 6.1+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...