2-(Trifluoromethoxy)anisole 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Methoxy-2-(trifluoromethoxy)benzene, 2-Trifluoromethoxyanisole

PC0515-25G PC0515-5G PC0515-1G
77412-614EA 138.5 USD
77412-614 77412-616 77412-612
2-(Trifluoromethoxy)anisole 97%
2-(Trifluoromethoxy)anisole
Formula: C₈H₇F₃O₂
MW: 192.14
Boiling Pt: 69…70 °C/10 mmHg
Flash Pt: 47 °C
Storage Temperature: Ambient
CAS Number: 261952-22-1
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR