4-Bromo-2-fluoroaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC1397E-100G PC1397E-25G
77413-150EA 28.52 USD
77413-150 77413-152
4-Bromo-2-fluoroaniline 98%
4-Bromo-2-fluoroaniline
Formula: C₆H₅BrFN
MW: 190.01 g/mol
Boiling Pt: 103…108 °C/13 mmHg
Melting Pt: 38…42 °C
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00010221
CAS Number: 367-24-8
EINECS: 206-685-4
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR