3-Fluoro-4-(trifluoromethoxy)phenylacetylene 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Ethynyl-2-fluoro-1-(trifluoromethoxy)benzene, 4-Ethynyl-α,α,α,2-tetrafluoroanisole

PC1161-1G PC1161-5G
77413-072EA 55.02 USD
77413-072 77413-074
3-Fluoro-4-(trifluoromethoxy)phenylacetylene 98%
3-Fluoro-4-(trifluoromethoxy)phenylacetylene
Formula: C₉H₄F₄O
MW: 204.121 g/mol
Density: 1.279 g/cm³ (25 °C)
Flash Pt: 40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08458006
CAS Number: 912617-68-6
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR