1,5-Bis(diphenylphosphino)pentane 97%

Supplier: APOLLO SCIENTIFIC

Synonyms: Pentane-1,3-diylbis(diphenylphosphine), Pentylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)pentane

OR315706-100G OR315706-10G OR315706-25G
77409-092EA 116.14 USD
77409-092 77409-094 77409-096
1,5-Bis(diphenylphosphino)pentane 97%
1,5-Bis(diphenylphosphino)pentane
Formula: C₂₉H₃₀P₂
MW: 440.5 g/mol
Boiling Pt: 230
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00003052
CAS Number: 27721-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR