1-Benzyl-3-aminopyrrolidine 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
3-Amino-1-benzylpyrrolidine, 1-Benzylpyrrolidin-3-amine
OR30970-100G
OR30970-25G
OR30970-5G
77408-996EA
716.22
USD
77408-996
77408-998
77409-000
1-Benzyl-3-aminopyrrolidine 97%
1-Benzyl-3-aminopyrrolidine
Formula:
C₁₁H₁₆N₂ MW: 176.26 g/mol Density: 1.024 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00059036 CAS Number: 18471-40-4 UN: 2735 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...