(5-Methylpyridin-2-yl)methanamine
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR306469-1G
OR306469-500MG
OR306469-5G
77408-922EA
162
USD
77408-922
77408-924
77408-926
(5-Methylpyridin-2-yl)methanamine
(5-Methylpyridin-2-yl)methanamine
Formula:
C₇H₁₀N₂ MW: 122.1677 g/mol Boiling Pt: 102…105 °C/12 mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD08752541 CAS Number: 45715-08-0 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...