N-Benzylacrylamide 95%

Supplier: APOLLO SCIENTIFIC

OR30622-1G OR30622-5G
77408-914EA 21.28 USD
77408-914 77408-916
N-Benzylacrylamide 95%
N-Benzylacrylamide
Formula: CH₂=CHCONHCH₂C₆H₅
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00015333
CAS Number: 13304-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR