Picolinic acid
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Pyridine-2-carboxylic acid, 2-Picolinic acid, 2-Pyridinecarboxylic acid
OR23054-500G
OR23054-5G
77407-046EA
83.54
USD
77407-046
77407-048
Picolinic acid
Picolinic acid
Formula:
C₆H₅NO₂ MW: 123.11 g/mol Melting Pt: 139…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006293 CAS Number: 98-98-6 EINECS: 202-719-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...