Tributyltin chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Chlorotributylstannane, Tri-n-butyltin chloride, TBTC

OR2288-250G OR2288-50G OR2288-5G
77406-968EA 130.4 USD
77406-968 77406-970 77406-972
Tributyltin chloride 95%
Tributyltin chloride
Formula: C₁₂H₂₇ClSn
MW: 325.51 g/mol
Boiling Pt: 171…173 °C/25 mmHg
Melting Pt: -9
Density: 1.186 g/cm³ (25 °C)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00000521
CAS Number: 1461-22-9
EINECS: 215-958-7
UN: 2788
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR