6-Hydroxy-m-toluic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Methylsalicylic acid, 2,5-Cresotic acid

OR28783-100G OR28783-25G
77408-236EA 81.5 USD
77408-236 77408-238
6-Hydroxy-m-toluic acid
6-Hydroxy-m-toluic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 313
Melting Pt: 152
Storage Temperature: Ambient
MDL Number: MFCD00002461
CAS Number: 89-56-5
EINECS: 201-918-6
Merck Index: 12,02650

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR