3-Bromothiophenol 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: m-Bromothiophenol, 3-Bromobenzenethiol, 1-Bromo-3-mercaptobenzene, 3-Bromobenzene-1-thiol

OR2454-1G OR2454-25G OR2454-5G
77407-496EA 22.42 USD
77407-496 77407-498 77407-500
3-Bromothiophenol 97%
3-Bromothiophenol
Formula: C₆H₅BrS
MW: 189.08 g/mol
Boiling Pt: 123…124 °C/40 mmHg
Density: 1.832 g/cm³ (25 °C)
Flash Pt: 105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009603
CAS Number: 6320-01-0
EINECS: 228-664-9
UN: 2922
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR