Iodoethane 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Ethyl iodide

OR16933-100G OR16933-500G
77405-850EA 83.54 USD
77405-850 77405-852
Iodoethane 98%
Iodoethane
Formula: C₂H₅I
MW: 155.97 g/mol
Boiling Pt: 72
Melting Pt: -108
Density: 1.95 g/cm³ (25 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00001091
CAS Number: 75-03-6
EINECS: 200-833-1
UN: 1993
ADR: 3 III
Merck Index: 13,03846

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR