trans-N1,N2-Dimethylcyclohexane-1,2-diamine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: trans-N,N'-Dimethyl-1,2-cyclohexanediamine

OR17015-1G OR17015-25G OR17015-5G
77405-910EA 21.26 USD
77405-910 77405-912 77405-914
trans-N1,N2-Dimethylcyclohexane-1,2-diamine
trans-N1,N2-Dimethylcyclohexane-1,2-diamine
Formula: C₈H₁₈N₂
MW: 142.24 g/mol
Boiling Pt: 78…80 °C/18 mmHg
Density: 0.902 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD03001702
CAS Number: 67579-81-1
UN: 2735
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR