(2R,3S)-3-Hydroxy-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)butanoic acid
Supplier: APOLLO SCIENTIFIC
OR110051-1G
OR110051-5G
77403-038EA
176.26
USD
77403-038
77403-040
(2R,3S)-3-Hydroxy-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)butanoic acid
(2R,3S)-3-Hydroxy-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)butanoic acid
Formula:
C₁₂H₁₃NO₄ MW: 235.236 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24850048 CAS Number: 1440531-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...