6-Chloro-N4-methylpyrimidine-2,4-diamine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Amino-4-chloro-6-(methylamino)pyrimidine, 2-Amino-4-chloro-6-(methylamino)-1,3-diazine

OR110703-10G OR110703-1G OR110703-5G
77403-432EA 274.58 USD
77403-432 77403-434 77403-436
6-Chloro-N4-methylpyrimidine-2,4-diamine
6-Chloro-N4-methylpyrimidine-2,4-diamine
Formula: C₅H₇ClN₄
MW: 158.589 g/mol
Melting Pt: 159…162 °C
Storage Temperature: Ambient
MDL Number: MFCD02091108
CAS Number: 1005-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR