Diethyl (hydroxymethyl)phosphonate 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR10715-100G OR10715-25G
77402-906EA 52.98 USD
77402-906 77402-908
Diethyl (hydroxymethyl)phosphonate 97%
Diethyl (hydroxymethyl)phosphonate
Formula: C₅H₁₃O₄P
Boiling Pt: 124…126 °C/3 mmHg
Density: 1.14 g/cm³ (25 °C)
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00014418
CAS Number: 3084-40-0
EINECS: 221-391-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR