N-Ethyl-4-pentylbenzamide

Supplier: APOLLO SCIENTIFIC

OR110453-1G OR110453-250MG OR110453-5G
77403-356EA 176.26 USD
77403-356 77403-358 77403-360
N-Ethyl-4-pentylbenzamide
N-Ethyl-4-pentylbenzamide
Formula: C₁₄H₂₁NO
MW: 219.323 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03551084
CAS Number: 401587-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR