1,2-Dimethylimidazole-4-sulfonyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1,2-Dimethyl-1H-imidazole-4-sulfonyl chloride, 1,2-Dimethyl-1H-imidazole-4-sulphonyl chloride

OR0412-1G OR0412-250MG
77402-062EA 68.9 USD
77402-062 77402-064
1,2-Dimethylimidazole-4-sulfonyl chloride 95%
1,2-Dimethylimidazole-4-sulphonyl chloride
Formula: C₅H₇ClN₂O₂S
Melting Pt: 91…94 °C
Storage Temperature: Ambient
MDL Number: MFCD01313780
CAS Number: 137049-02-6
UN: 1759
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR