2-Chloroethylammonium chloride 97%

Supplier: APOLLO SCIENTIFIC

Synonyms: 2-Chloroethylamine hydrochloride, 2-Chloroethylamine HCl, 2-Chloroethanamine hydrochloride

OR0986-100G OR0986-25G
77402-368EA 35.68 USD
77402-368 77402-370
2-Chloroethylammonium chloride 97%
2-Chloroethylammonium chloride
Formula: C₂H₇Cl₂N
MW: 115.99 g/mol
Melting Pt: 140…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00012887
CAS Number: 870-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR