3,5-Dichlorophenol
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR0059-100G
OR0059-25G
OR0059-5G
77401-668EA
85.58
USD
77401-668
77401-670
77401-672
3,5-Dichlorophenol
3,5-Dichlorophenol
Formula:
C₆H₄Cl₂O Boiling Pt: 233 °C Melting Pt: 63…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002259 CAS Number: 591-35-5 EINECS: 209-714-9 UN: 2020 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...